CAS RN 263717-53-9
CAS RN 263717-53-9
IUPAC Name:
4-[1,5-bis(4-hydroxyphenyl)-4-propyl-2,3-dihydro-1H-pyrazol-3-ylidene]cyclohexa-2,5-dien-1-one
Synonyme:
propylpyrazoletriol
Molekulargewicht (g/mol):
386.45
Summenformel:
C24H22N2O3
InChi Key:
UOSWGERPQQOSHS-UHFFFAOYSA-N
SMILES:
CCCC1=C(N(NC1=C1C=CC(=O)C=C1)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
1
–
2
von
2
Ergebnisse
1
Tocris Bioscience™ PPT
CAS: 263717-53-9 Summenformel: C24H22N2O3
2
Propyl pyrazole triol, MedChemExpress
MedChemExpress Propyl pyrazole triol (PPT) is a selective estrogen receptor alpha (ERα) agonist. The relative binding affinity of Propyl pyrazole triol for ERα (ERα: 49%) around 410 times higher compared with estrogen receptor beta (ERβ: 0.12%).