CAS RN 30578-37-1
IUPAC Name:
4-amino-6-methoxy-1-phenyl-1λ⁵-pyridazin-1-ylium methyl sulfate
Synonyme:
amezinium metilsulfate 甲硫阿美铵
Molekulargewicht (g/mol):
313.33
Summenformel:
C12H15N3O5S
InChi Key:
ZEASXVYVFFXULL-UHFFFAOYSA-N
SMILES:
COS([O-])(=O)=O.COC1=CC(N)=CN=[N+]1C1=CC=CC=C1
1
–
1
von
1
Ergebnisse
1
Amezinium methylsulfate, MedChemExpress
MedChemExpress Amezinium metilsulfate has multiple mechanisms, including stimulation of alpha and beta-1 receptors and inhibition ofnoradrenaline and tyramine uptake.