CAS RN 328968-36-1
CAS RN 328968-36-1
IUPAC Name:
4-(4-{[5-(4,5-dimethyl-2-nitrophenyl)furan-2-yl]methylidene}-3-methyl-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzoic acid
Synonyme:
4-(4-{[5-(4,5-dimethyl-2-nitrophenyl)furan-2-yl]methylidene}-3-methyl-5-oxopyrazol-1-yl)benzoic acid
Molekulargewicht (g/mol):
445.43
Summenformel:
C24H19N3O6
InChi Key:
HEKJYZZSCQBJGB-UHFFFAOYSA-N
SMILES:
CC1=NN(C(=O)C1=CC1=CC=C(O1)C1=CC(C)=C(C)C=C1[N+]([O-])=O)C1=CC=C(C=C1)C(O)=O
1
–
2
von
2
Ergebnisse
1
C646, MedChemExpress
MedChemExpress C646 is a selective and competitive histone acetyltransferase p300 inhibitor with Ki of 400 nM, and is less potent for other acetyltransferases.
2
C 646, Tocris Bioscience™
CAS: 328968-36-1 Summenformel: C24H19N3O6 Molekulargewicht (g/mol): 445.43 MDL-Nummer: MFCD01784780 InChI-Schlüssel: HEKJYZZSCQBJGB-UHFFFAOYSA-N Synonym: 4-4-5-4,5-dimethyl-2-nitrophenyl furan-2-yl methylidene-3-methyl-5-oxopyrazol-1-yl benzoic acid PubChem CID: 2871948 IUPAC-Name: 4-(4-{[5-(4,5-dimethyl-2-nitrophenyl)furan-2-yl]methylidene}-3-methyl-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzoic acid SMILES: CC1=NN(C(=O)C1=CC1=CC=C(O1)C1=CC(C)=C(C)C=C1[N+]([O-])=O)C1=CC=C(C=C1)C(O)=O