CAS RN 349-59-7
CAS RN 349-59-7
IUPAC Name:
1-ethenyl-3,5-bis(trifluoromethyl)benzene
Synonyme:
1-ethenyl-3,5-bis(trifluoromethyl)benzene
Molekulargewicht (g/mol):
240.15
Summenformel:
C10H6F6
InChi Key:
LFICVUCVPKKPFF-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC(=CC(C=C)=C1)C(F)(F)F
1
–
1
von
1
Ergebnisse
1
3,5-Bis(trifluormethyl)styrol, 98 %, stab., Thermo Scientific Chemicals
CAS: 349-59-7 Summenformel: C10H6F6 Molekulargewicht (g/mol): 240.15 MDL-Nummer: MFCD00075538 InChI-Schlüssel: LFICVUCVPKKPFF-UHFFFAOYSA-N Synonym: 3,5-bis trifluoromethyl styrene,1,3-bis trifluoromethyl-5-vinylbenzene,1-ethenyl-3,5-bis trifluoromethyl benzene,3,5-bis trifluoromethyl styrene, stab. with 4-methoxyphenol,pubchem2736,acmc-1cmtq,3,5-bis trifluromethyl styrene,1,3-bis-trifluoromethyl-5-vinyl-benzene,3,5-bis trifluoromethyl styrene, stab. PubChem CID: 2773242 IUPAC-Name: 1-ethenyl-3,5-bis(trifluormethyl)benzol SMILES: FC(F)(F)C1=CC(=CC(C=C)=C1)C(F)(F)F