CAS RN 61-73-4
IUPAC Name:
3,7-bis(dimethylamino)-5λ⁴-phenothiazin-5-ylium chloride
Synonyme:
methylene blue methylthioninium chloride methylene blue anhydrous methylthioninii chloridum azul de metileno bleu de methylene cloruro de metiltioninio methylenblau chlorure de methylthioninium basic blue 9
Molekulargewicht (g/mol):
319.85
Summenformel:
C16H18ClN3S
InChi Key:
CXKWCBBOMKCUKX-UHFFFAOYSA-M
SMILES:
[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
1
–
4
von
4
Ergebnisse
3
MedChemExpress Methylene Blue
MedChemExpress Methylene blue (Basic Blue 9) is a guanylyl cyclase (sGC), monoamine oxidase A (MAO-A) and NO synthase (NOS) inhibitor.