CAS RN 790186-68-4
CAS RN 790186-68-4
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
GW284543, MedChemExpress
MedChemExpress GW284543 (UNC10225170) is a selective MEK5 inhibitor. GW284543 (UNC10225170) reduces pERK5, and decreases endogenous MYC protein.