CAS RN 900-91-4
CAS RN 900-91-4
IUPAC Name:
3,3,3-triphenylpropanoic acid
Synonyme:
3,3,3-triphenylpropanoic acid
Molekulargewicht (g/mol):
302.37
Summenformel:
C21H18O2
InChi Key:
XMSJLUKCGWQAHO-UHFFFAOYSA-N
SMILES:
OC(=O)CC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
von
1
Ergebnisse
1
3,3,3-Triphenylpropionsäure, 98 %, Thermo Scientific™
CAS: 900-91-4 Summenformel: C21H18O2 Molekulargewicht (g/mol): 302.373 MDL-Nummer: MFCD00002713 InChI-Schlüssel: XMSJLUKCGWQAHO-UHFFFAOYSA-N Synonym: 3,3,3-triphenylpropionic acid,tritylacetic acid,3,3,3-triphenyl propanoic acid,2-tritylacetic acid,pubchem12502,acmc-209r4v,2-triphenylmethylacetic acid,2-triphenylmethyl acetic acid,3,3 3-triphenyl propionic acid PubChem CID: 136696 IUPAC-Name: 3,3,3-triphenylpropansäure SMILES: C1=CC=C(C=C1)C(CC(=O)O)(C2=CC=CC=C2)C3=CC=CC=C3