CAS RN 96487-37-5
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Nuvenzepine, MedChemExpress
MedChemExpress Nuvenzepine is an mAChR antagonist, has the potential for gastrospasm treatment.