CAS RN 1310-66-3
CAS RN 1310-66-3
Lithiumhydroxid-Monohydrat, 98+ %, ACS-Reagenz, Thermo Scientific Chemicals
CAS: 1310-66-3 Summenformel: H3LiO2 Molekulargewicht (g/mol): 41.96 MDL-Nummer: MFCD00149772 InChI-Schlüssel: GLXDVVHUTZTUQK-UHFFFAOYSA-M Synonym: lithium hydroxide monohydrate,lithium hydroxide hydrate,lithium hydroxido,hydroxyde de lithium,unii-g51xlp968g,lithium hydroxide, monohydrate,lithiumhydrate,lioh-hydrate,lioh.hydrate,lioh water PubChem CID: 168937 SMILES: [Li+].O.[OH-]
Lithiumhydroxid-Monohydrat, reinst, 56 % LiOH, Thermo Scientific Chemicals
CAS: 1310-66-3 Summenformel: H3LiO2 Molekulargewicht (g/mol): 41.96 MDL-Nummer: MFCD00149772 InChI-Schlüssel: GLXDVVHUTZTUQK-UHFFFAOYSA-M Synonym: lithium hydroxide monohydrate,lithium hydroxide hydrate,lithium hydroxido,hydroxyde de lithium,unii-g51xlp968g,lithium hydroxide, monohydrate,lithiumhydrate,lioh-hydrate,lioh.hydrate,lioh water PubChem CID: 168937 SMILES: [Li+].O.[OH-]
Lithiumhydroxid-Monohydrat, 56.5 % min., Thermo Scientific Chemicals
CAS: 1310-66-3 Summenformel: H3LiO2 Molekulargewicht (g/mol): 41.96 MDL-Nummer: MFCD00149772 InChI-Schlüssel: GLXDVVHUTZTUQK-UHFFFAOYSA-M Synonym: lithium hydroxide monohydrate,lithium hydroxide hydrate,lithium hydroxido,hydroxyde de lithium,unii-g51xlp968g,lithium hydroxide, monohydrate,lithiumhydrate,lioh-hydrate,lioh.hydrate,lioh water PubChem CID: 168937 SMILES: [Li+].O.[OH-]
Lithiumhydroxidmonohydrat, reinst, SLR, Fisher Chemical
CAS: 1310-66-3 Summenformel: H3LiO2 Molekulargewicht (g/mol): 41.96 MDL-Nummer: MFCD00149772 InChI-Schlüssel: GLXDVVHUTZTUQK-UHFFFAOYSA-M Synonym: lithium hydroxide monohydrate,lithium hydroxide hydrate,lithium hydroxido,hydroxyde de lithium,unii-g51xlp968g,lithium hydroxide, monohydrate,lithiumhydrate,lioh-hydrate,lioh.hydrate,lioh water PubChem CID: 168937 IUPAC-Name: Lithium(1+)-Hydrat-Hydroxid SMILES: [Li+].O.[OH-]
Lithiumhydroxid-Monohydrat, ACS, 98 % min., Thermo Scientific Chemicals
CAS: 1310-66-3 Summenformel: H3LiO2 Molekulargewicht (g/mol): 41.96 MDL-Nummer: MFCD00149772 InChI-Schlüssel: GLXDVVHUTZTUQK-UHFFFAOYSA-M Synonym: lithium hydroxide monohydrate,lithium hydroxide hydrate,lithium hydroxido,hydroxyde de lithium,unii-g51xlp968g,lithium hydroxide, monohydrate,lithiumhydrate,lioh-hydrate,lioh.hydrate,lioh water PubChem CID: 168937 SMILES: [Li+].O.[OH-]
Lithiumhydroxid-Monohydrat, weißes, kristallines Pulver, 99.995 % (Metallbasis), Thermo Scientific Chemicals
CAS: 1310-66-3 Summenformel: H3LiO2 Molekulargewicht (g/mol): 41.96 MDL-Nummer: MFCD00149772 InChI-Schlüssel: GLXDVVHUTZTUQK-UHFFFAOYSA-M Synonym: lithium hydroxide monohydrate,lithium hydroxide hydrate,lithium hydroxido,hydroxyde de lithium,unii-g51xlp968g,lithium hydroxide, monohydrate,lithiumhydrate,lioh-hydrate,lioh.hydrate,lioh water PubChem CID: 168937 IUPAC-Name: Lithium;hydroxid;hydrat SMILES: [Li+].O.[OH-]
Lithium Hydroxide Monohydrate, TRC
CAS: 1310-66-3 Summenformel: HLiO.H2O Molekulargewicht (g/mol): 42 Synonym: Lithium Hydroxide Hydrate; IUPAC-Name: hydroxylithium;hydrate SMILES: O.[Li]O
Lithiumhydroxid-Monohydrat, Batterie-Gütegrad, Thermo Scientific Chemicals
CAS: 1310-66-3 Summenformel: H3LiO2 Molekulargewicht (g/mol): 41.96 MDL-Nummer: MFCD00149772 InChI-Schlüssel: GLXDVVHUTZTUQK-UHFFFAOYSA-M Synonym: lithium hydroxide monohydrate,lithium hydroxide hydrate,lithium hydroxido,hydroxyde de lithium,unii-g51xlp968g,lithium hydroxide, monohydrate,lithiumhydrate,lioh-hydrate,lioh.hydrate,lioh water PubChem CID: 168937 IUPAC-Name: Lithium;hydroxid;hydrat SMILES: [Li+].O.[OH-]
Lithiumhydroxidmonohydrat, 98 %, Thermo Scientific Chemicals
CAS: 1310-66-3 Summenformel: H3LiO2 Molekulargewicht (g/mol): 41.96 MDL-Nummer: MFCD00149772 InChI-Schlüssel: GLXDVVHUTZTUQK-UHFFFAOYSA-M Synonym: lithium hydroxide monohydrate,lithium hydroxide hydrate,lithium hydroxido,hydroxyde de lithium,unii-g51xlp968g,lithium hydroxide, monohydrate,lithiumhydrate,lioh-hydrate,lioh.hydrate,lioh water PubChem CID: 168937 IUPAC-Name: Lithium;hydroxid;hydrat SMILES: [Li+].O.[OH-]
Lithiumhydroxid-Monohydrat, Honeywell Fluka™
CAS: 1310-66-3 Summenformel: H3LiO2 Molekulargewicht (g/mol): 41.96 MDL-Nummer: MFCD00149772 InChI-Schlüssel: GLXDVVHUTZTUQK-UHFFFAOYSA-M Synonym: Lithiumhydroxid-Monohydrat,Lithiumhydroxid-Hydrat,Lithiumhydroxid,Lithiumhydroxyd,Lithiumhydroxid, Monohydrat,Lithiumhydrat,LiOH-Hydrat,LiOH.Hydrat,LiOH Wasser,lioh water PubChem CID: 168937 SMILES: [Li+].O.[OH-]
Lithiumhydroxid-Monohydrat, Honeywell Fluka™
CAS: 1310-66-3 Summenformel: H3LiO2 Molekulargewicht (g/mol): 41.96 MDL-Nummer: MFCD00149772 InChI-Schlüssel: GLXDVVHUTZTUQK-UHFFFAOYSA-M Synonym: Lithiumhydroxid-Monohydrat,Lithiumhydroxid-Hydrat,Lithiumhydroxid,Lithiumhydroxyd,Lithiumhydroxid, Monohydrat,Lithiumhydrat,LiOH-Hydrat,LiOH.Hydrat,LiOH Wasser,lioh water PubChem CID: 168937 IUPAC-Name: Lithium;hydroxid;hydrat SMILES: [Li+].O.[OH-]