CAS RN 6381-59-5
Kalium-Natriumtartrat-Tetrahydrat, ACS-Reagenz, Thermo Scientific Chemicals
CAS: 6381-59-5 Summenformel: C4H4KNaO6·4H2O Molekulargewicht (g/mol): 282.22 InChI-Schlüssel: VZOPRCCTKLAGPN-UHFFFAOYSA-L Synonym: potassium sodium tartrate tetrahydrate,sodium potassium tartrate tetrahydrate,monopotassium monosodium tartrate tetrahydrate,sodiumpotassiumtartrate,tetr,potassium sodium 2,3-dihydroxybutanedioate tetrahydrate,potassium sodium tetrahydrate tartrate,l +-tartaric acid potassium sodium salt,acmc-209nif,acmc-20aju8,ksc493o7j PubChem CID: 2724148 IUPAC-Name: Kalium; Natrium; 2,3-Dihydroxybutanandioat; Tetrahydrat SMILES: C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
Kalium-Natriumtartrat-Tetrahydrat, +99%, zur Analyse, Thermo Scientific Chemicals
CAS: 6381-59-5 MDL-Nummer: MFCD00150989 InChI-Schlüssel: VZOPRCCTKLAGPN-UHFFFAOYSA-L Synonym: potassium sodium tartrate tetrahydrate,sodium potassium tartrate tetrahydrate,monopotassium monosodium tartrate tetrahydrate,sodiumpotassiumtartrate,tetr,potassium sodium 2,3-dihydroxybutanedioate tetrahydrate,potassium sodium tetrahydrate tartrate,l +-tartaric acid potassium sodium salt,acmc-209nif,acmc-20aju8,ksc493o7j PubChem CID: 2724148 IUPAC-Name: Kalium; Natrium; 2,3-Dihydroxybutanandioat; Tetrahydrat SMILES: C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
Kaliumnatriumtartrat-Tetrahydrat, AR-zertifiziert für die Analyse, Fisher Chemical™
CAS: 6381-59-5 Summenformel: C4H12KNaO10 Molekulargewicht (g/mol): 282.218 MDL-Nummer: 150989 InChI-Schlüssel: VZOPRCCTKLAGPN-UHFFFAOYSA-L Synonym: potassium sodium tartrate tetrahydrate,sodium potassium tartrate tetrahydrate,monopotassium monosodium tartrate tetrahydrate,sodiumpotassiumtartrate,tetr,potassium sodium 2,3-dihydroxybutanedioate tetrahydrate,potassium sodium tetrahydrate tartrate,l +-tartaric acid potassium sodium salt,acmc-209nif,acmc-20aju8,ksc493o7j PubChem CID: 2724148 IUPAC-Name: Kalium; Natrium; 2,3-Dihydroxybutanandioat; Tetrahydrat SMILES: C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
Kaliumnatrium L-Tartrat-Tetrahydrat, 99%, Thermo Scientific Chemicals
CAS: 6381-59-5 Summenformel: C4H12KNaO10 Molekulargewicht (g/mol): 282.218 MDL-Nummer: MFCD00150989 InChI-Schlüssel: VZOPRCCTKLAGPN-UHFFFAOYSA-L Synonym: potassium sodium tartrate tetrahydrate,sodium potassium tartrate tetrahydrate,monopotassium monosodium tartrate tetrahydrate,sodiumpotassiumtartrate,tetr,potassium sodium 2,3-dihydroxybutanedioate tetrahydrate,potassium sodium tetrahydrate tartrate,l +-tartaric acid potassium sodium salt,acmc-209nif,acmc-20aju8,ksc493o7j PubChem CID: 2724148 IUPAC-Name: Kalium; Natrium; 2,3-Dihydroxybutanandioat; Tetrahydrat SMILES: C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
Kaliumnatrium-L-Tartrat-Tetrahydrat, ACS, 99.0 bis 102.0 %, Thermo Scientific Chemicals
CAS: 6381-59-5 Summenformel: C4H12KNaO10 Molekulargewicht (g/mol): 282.218 MDL-Nummer: MFCD00150989 InChI-Schlüssel: VZOPRCCTKLAGPN-UHFFFAOYSA-L Synonym: potassium sodium tartrate tetrahydrate,sodium potassium tartrate tetrahydrate,monopotassium monosodium tartrate tetrahydrate,sodiumpotassiumtartrate,tetr,potassium sodium 2,3-dihydroxybutanedioate tetrahydrate,potassium sodium tetrahydrate tartrate,l +-tartaric acid potassium sodium salt,acmc-209nif,acmc-20aju8,ksc493o7j PubChem CID: 2724148 IUPAC-Name: Kalium; Natrium; 2,3-Dihydroxybutanandioat; Tetrahydrat SMILES: C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
Kalium-Natriumtartrat-Tetrahydrat, 99.0 bis 102.0 %, (ACS-Reagenzienqualität), MP Biomedicals
CAS: 6381-59-5 Summenformel: C4H12KNaO10 Molekulargewicht (g/mol): 282.218 InChI-Schlüssel: VZOPRCCTKLAGPN-UHFFFAOYSA-L Synonym: Kaliumnatriumtartrat-Tetrahydrat,Natrium-Kalium-Tartrat-Tetrahydrat,Monokaliummononatriumtartrat-Tetrahydrat,Natrium-Kalium-Tartrat, Tetr,Kaliumnatrium2,3-dihydroxybutanedioat-Tetrahydrat,Kaliumnatriumtetrahydrat-Tartrat,L+-Weinsäure-Kaliumnatriumsalz,acmc-209nif,acmc-20aju8,ksc493o7j PubChem CID: 2724148 IUPAC-Name: Kalium; Natrium; 2,3-Dihydroxybutanandioat; Tetrahydrat SMILES: C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
Natrium-Kalium-Tartrat-Tetrahydrat,≥ 99 %, MP Biomedicals™
CAS: 6381-59-5 Summenformel: C4H12KNaO10 MDL-Nummer: MFCD00150989 Synonym: Kaliumnatriumtartrat-Tetrahydrat,Natrium-Kalium-Tartrat-Tetrahydrat,Monokaliummononatriumtartrat-Tetrahydrat,Natrium-Kalium-Tartrat, Tetr,Kaliumnatrium2,3-dihydroxybutanedioat-Tetrahydrat,Kaliumnatriumtetrahydrat-Tartrat,L+-Weinsäure-Kaliumnatriumsalz,acmc-209nif,acmc-20aju8,ksc493o7j PubChem CID: 2724148
Kaliumnatriumtartrat-Tetrahydrat, analytische Qualität, Penta Chemicals
CAS: 6381-59-5 Molekulargewicht (g/mol): 282.23
Rochelle's Salt, TRC
Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.
Rochelle's Salt, TRC
Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.
Rochelle's Salt, TRC
Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.
- 1
- 2