CAS RN 88495-63-0
CAS RN 88495-63-0
IUPAC Name:
4-oxo-4-{[(4S,5R,8S,9R,10R,12R,13R)-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo[10.3.1.0⁴,¹³.0⁸,¹³]hexadecan-10-yl]oxy}butanoic acid
Synonyme:
artesunate
Molekulargewicht (g/mol):
384.43
Summenformel:
C19H28O8
InChi Key:
FIHJKUPKCHIPAT-JQXDXKTESA-N
SMILES:
C[C@@H]1CC[C@H]2[C@@H](C)[C@@H](OC(=O)CCC(O)=O)O[C@@H]3OC4(C)CC[C@@H]1[C@@]23OO4
1
–
4
von
4
Ergebnisse
1
Artesunat, Thermo Scientific Chemicals
CAS: 88495-63-0 Summenformel: C19H28O8 Molekulargewicht (g/mol): 384.42 InChI-Schlüssel: FIHJKUPKCHIPAT-RVBFJZMGSA-N Synonym: artesunate PubChem CID: 129317578 SMILES: CC1CCC2C(C(OC3C24C1CCC(O3)(OO4)C)OC(=O)CCC(=O)O)C
2
Artesunate, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Artesunate, MedChemExpress
MedChemExpress Artesunate is an inhibitor of both STAT-3 and exported protein 1 (EXP1).
4
Tocris Bioscience™ Artesunate
Suppresses ROS-induced activation of the NLRP3 inflammasome