CAS RN 101827-46-7
CAS RN 101827-46-7
IUPAC Name:
hydrogen [(4-tert-butylphenyl)methyl](methyl)[(naphthalen-1-yl)methyl]amine chloride
Synonyme:
hydrogen butenafine chloride
Molekulargewicht (g/mol):
353.93
Summenformel:
C23H28ClN
InChi Key:
LJBSAUIFGPSHCN-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CN(CC1=CC=C(C=C1)C(C)(C)C)CC1=C2C=CC=CC2=CC=C1
1
–
1
von
1
Ergebnisse
1
Butenafine Hydrochloride, MedChemExpress
MedChemExpress Butenafine Hydrochloride (KP363 Hydrochloride) is a synthetic benzylamine antifungal, works by inhibiting the synthesis of sterols by inhibiting squalene epoxidase.
