CAS RN 87-89-8
Thermo Scientific Chemicals Myo-Inositol, +98 %
CAS: 87-89-8 Summenformel: C6H12O6. Molekulargewicht (g/mol): 180.156 MDL-Nummer: MFCD00077932 InChI-Schlüssel: CDAISMWEOUEBRE-UHFFFAOYSA-N Synonym: scyllo-inositol,myo-inositol,inositol,muco-inositol,epi-inositol,i-inositol,meso-inositol,allo-inositol,1d-chiro-inositol,myoinositol PubChem CID: 892 ChEBI: CHEBI:24848 IUPAC-Name: Cyclohexan-1,2,3,4,5,6-Hexol SMILES: C1(C(C(C(C(C1O)O)O)O)O)O
Thermo Scientific Chemicals Myo-Inositol, Zellkulturqualität
CAS: 87-89-8 Summenformel: C6H12O6. Molekulargewicht (g/mol): 180.156 MDL-Nummer: MFCD00077932 InChI-Schlüssel: CDAISMWEOUEBRE-UHFFFAOYSA-N Synonym: scyllo-inositol,myo-inositol,inositol,muco-inositol,epi-inositol,i-inositol,meso-inositol,allo-inositol,1d-chiro-inositol,myoinositol PubChem CID: 892 ChEBI: CHEBI:24848 IUPAC-Name: Cyclohexan-1,2,3,4,5,6-Hexol SMILES: C1(C(C(C(C(C1O)O)O)O)O)O
Thermo Scientific Chemicals Inositol, 98+%
CAS: 87-89-8 Summenformel: C6H12O6 Molekulargewicht (g/mol): 180.16 MDL-Nummer: MFCD00077932 InChI-Schlüssel: CDAISMWEOUEBRE-UHFFFAOYSA-N Synonym: scyllo-inositol,myo-inositol,inositol,muco-inositol,epi-inositol,i-inositol,meso-inositol,allo-inositol,1d-chiro-inositol,myoinositol PubChem CID: 892 ChEBI: CHEBI:24848 IUPAC-Name: Cyclohexan-1,2,3,4,5,6-Hexol SMILES: C1(C(C(C(C(C1O)O)O)O)O)O
myo-Inositol, 99%, Thermo Scientific Chemicals
CAS: 87-89-8 Summenformel: C6H12O6. Molekulargewicht (g/mol): 180.156 MDL-Nummer: MFCD00077932 InChI-Schlüssel: CDAISMWEOUEBRE-UHFFFAOYSA-N Synonym: scyllo-inositol,myo-inositol,inositol,muco-inositol,epi-inositol,i-inositol,meso-inositol,allo-inositol,1d-chiro-inositol,myoinositol PubChem CID: 892 ChEBI: CHEBI:24848 IUPAC-Name: Cyclohexan-1,2,3,4,5,6-Hexol SMILES: C1(C(C(C(C(C1O)O)O)O)O)O
D-myo-Inositol, 99,2 %, Zellkulturreagenz, für HPLC-Analysen MP Biomedicals™
CAS: 87-89-8 Summenformel: C6H12O6. Molekulargewicht (g/mol): 180.156 MDL-Nummer: MFCD00077932 InChI-Schlüssel: CDAISMWEOUEBRE-UHFFFAOYSA-N Synonym: scyllo-inositol,myo-inositol,inositol,muco-inositol,epi-inositol,i-inositol,meso-inositol,allo-inositol,1d-chiro-inositol,myoinositol PubChem CID: 892 ChEBI: CHEBI:24848 IUPAC-Name: Cyclohexan-1,2,3,4,5,6-Hexol SMILES: C1(C(C(C(C(C1O)O)O)O)O)O
D-Myo-inositol, 99.26 %, für HPLC-Analysen, MP Biomedicals™
CAS: 87-89-8 Summenformel: C6H12O6. Molekulargewicht (g/mol): 180.156 InChI-Schlüssel: CDAISMWEOUEBRE-UHFFFAOYSA-N Synonym: scyllo-inositol,myo-inositol,inositol,muco-inositol,epi-inositol,i-inositol,meso-inositol,allo-inositol,1d-chiro-inositol,myoinositol PubChem CID: 892 ChEBI: CHEBI:24848 IUPAC-Name: Cyclohexan-1,2,3,4,5,6-Hexol SMILES: C1(C(C(C(C(C1O)O)O)O)O)O
Myo-Inositol, TRC
CAS: 87-89-8 Summenformel: C6 H12 O6 Molekulargewicht (g/mol): 180.16 Synonym: myo-Inositol,Inositol, myo- (8CI),Bios I,Cyclohexanehexol,Cyclohexitol,Dambose,Inosital,Inosite,Inositene,Inositina,Inositol,MI,Meat sugar,Mesoinosit,Mesoinosite,Mesoinositol,Mesol,Mesovit,Mouse antialopecia factor,Myoinosite,Myoinositol,Nucite,Phaseomannite,Phaseomannitol,Rat antispectacled eye factor,Scyllite,cis-1,2,3,5-trans-4,6-Cyclohexanehexol,i-Inositol,iso-Inositol,meso-Inositol IUPAC-Name: Cyclohexan-1,2,3,4,5,6-Hexol SMILES: O[C@@H]1[C@@H](O)[C@H](O)[C@H](O)[C@H](O)[C@H]1O
i-Inositol, MedChemExpress
MedChemExpress i-Inositol is a chemical compound, associated lipids are found in many foods, in particular fruit, especially cantaloupe and oranges.